Gene Transcripts

Gene Symbol
Amdhd1
Entrez Gene ID
71761
Gene Name
amidohydrolase domain containing 1
Gene Aliases
1300019J08Rik
Chromosome Location
Chr. 10: 93523338 - 93540043 on Build GRCm38
UniGene ID
Mm.244518
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027908.1 NP_082184.1 9 2327
GenBank mRNA AK090143.1 - 9 2319

View Full Product Details