Gene Transcripts

Gene Symbol
Pdilt
Entrez Gene ID
71830
Gene Name
protein disulfide isomerase-like, testis expressed
Gene Aliases
1700007B13Rik
Chromosome Location
Chr. 7: 119486586 - 119528621 on Build GRCm38
UniGene ID
Mm.84631
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027943.1 NP_082219.1 9 1431
GenBank mRNA AK005692.1 - 9 1431

View Full Product Details