Gene Transcripts
- Gene Symbol
- Haus5
- Entrez Gene ID
- 71909
- Gene Name
- HAUS augmin-like complex, subunit 5
- Gene Aliases
- 2310022K01Rik
- Chromosome Location
- Chr. 7: 30653708 - 30664994 on Build GRCm38
- UniGene ID
- Mm.290410
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027999.1 |
NP_082275.1
|
|
2187 |
|
View Full Product Details