Gene Transcripts

Gene Symbol
Klhl35
Entrez Gene ID
72184
Gene Name
kelch-like 35
Gene Aliases
2810406K13Rik
Chromosome Location
Chr. 7: 99464216 - 99474022 on Build GRCm38
UniGene ID
Mm.153585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028145.1 NP_082421.1 2, 3 964
GenBank mRNA AK013012.1 - 2, 3 942
BE994913.1 - 1, 2 369

View Full Product Details