Gene Transcripts

Gene Symbol
Serp2
Entrez Gene ID
72661
Gene Name
stress-associated endoplasmic reticulum protein family member 2
Gene Aliases
2810032E02Rik
Chromosome Location
Chr. 14: 76532812 - 76557919 on Build GRCm38
UniGene ID
Mm.320384
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160326.1 NP_001153798.1 3 733
GenBank mRNA AK012850.1 - 2 676
AY425619.1 - 3 718
BC100483.1 - 5 837
CF728547.1 - 2 492

View Full Product Details