Gene Transcripts
- Gene Symbol
- Hspb11
- Entrez Gene ID
- 72938
- Gene Name
- heat shock protein family B (small), member 11
- Gene Aliases
- 2900042B11Rik, IFT25
- Chromosome Location
- Chr. 4: 107253598 - 107284497 on Build GRCm38
- UniGene ID
- Mm.21787
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028394.2 |
NP_082670.1
|
3, 4 |
293 |
|
GenBank mRNA |
AK157651.1 |
- |
3, 4 |
294 |
|
View Full Product Details