Gene Transcripts

Gene Symbol
Arhgap36
Entrez Gene ID
75404
Gene Name
Rho GTPase activating protein 36
Gene Aliases
1100001E04Rik, rho-gap
Chromosome Location
Chr. X: 49448328 - 49500250 on Build GRCm38
UniGene ID
Mm.44305
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081123.1 NP_001074592.1 10, 11 1508
GenBank mRNA AB073967.1 - 10, 11 1375
AK048167.1 - 10, 11 1397
BC090672.1 - 10, 11 1479 30941825
BC145645.1 - 10, 11 1474 40130948

View Full Product Details