Gene Transcripts

Gene Symbol
Cypt12
Entrez Gene ID
75439
Gene Name
cysteine-rich perinuclear theca 12
Gene Aliases
1700001F04Rik, AV209098
Chromosome Location
Chr. 3: 17948444 - 17948952 on Build GRCm38
UniGene ID
Mm.27023
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029289.1 NP_083565.1 1 242
GenBank mRNA AK005586.1 - 1 243
AM040460.1 - 1 26

View Full Product Details