Gene Transcripts

Gene Symbol
Trdn
Entrez Gene ID
76757
Gene Name
triadin
Gene Aliases
2310045H21Rik, EG432451, TDN
Chromosome Location
Chr. 10: 33083483 - 33476709 on Build GRCm38
UniGene ID
Mm.338508
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029726.2 NP_084002.2 38 4002
GenBank mRNA AK036470.1 - 20 2542
AK085836.1 - 1 1627

View Full Product Details