Gene Transcripts

Gene Symbol
Hyls1
Entrez Gene ID
76832
Gene Name
hydrolethalus syndrome 1
Gene Aliases
3010015K02Rik
Chromosome Location
Chr. 9: 35560821 - 35570069 on Build GRCm38
UniGene ID
Mm.482285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029762.1 NP_084038.1 4 1130
GenBank mRNA AK013896.1 - 4 1164
AK131639.1 - 4 1094
BC035953.1 - 4 1106 4911163

View Full Product Details