Gene Transcripts

Gene Symbol
Axdnd1
Entrez Gene ID
77352
Gene Name
axonemal dynein light chain domain containing 1
Gene Aliases
9430070O13Rik, Gm979
Chromosome Location
Chr. 1: 156330415 - 156420915 on Build GRCm38
UniGene ID
Mm.428709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK134071.1 - 11 1374
BC150932.1 - 10 1226 9087843
BC172088.1 - 10 1316 9054374

View Full Product Details