Gene Transcripts

Gene Symbol
Iqch
Entrez Gene ID
78250
Gene Name
IQ motif containing H
Gene Aliases
4921504K03Rik
Chromosome Location
Chr. 9: 63421015 - 63602547 on Build GRCm38
UniGene ID
Mm.444612
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030068.1 NP_084344.1 17 2423
GenBank mRNA AK019535.1 - 17 2455
BC141047.1 - 17 2423 9055659
BC145231.1 - 17 2423 9053783

View Full Product Details