Gene Transcripts

Gene Symbol
Cypt15
Entrez Gene ID
78631
Gene Name
cysteine-rich perinuclear theca 15
Gene Aliases
1700085A12Rik
Chromosome Location
Chr. X: 39346266 - 39346963 on Build GRCm38
UniGene ID
Mm.243738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177380.1 NP_001170851.1 2 304
GenBank mRNA AK018908.1 - 2 304
BC100466.1 - 2 304

View Full Product Details