Gene Transcripts

Gene Symbol
Prg4
Entrez Gene ID
96875
Gene Name
proteoglycan 4 (megakaryocyte stimulating factor, articular superficial zone protein)
Gene Aliases
CACP, DOL54, JCAP, MSF, SZP, lubricin
Chromosome Location
Chr. 1: 150449412 - 150466221 on Build GRCm38
UniGene ID
Mm.174256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021400.3 NP_067375.3 7 1044
GenBank mRNA AB034730.1 - 7 1044

View Full Product Details