Gene Transcripts

Gene Symbol
Slk
Entrez Gene ID
54308
Gene Name
STE20-like kinase
Gene Aliases
Stk2
Chromosome Location
Chr. 1: 267359769 - 267415735 on Build Rnor_6.0
UniGene ID
Rn.228809
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019349.2 NP_062222.2 1 269
XM_006231558.2 XP_006231620.1 1 307

View Full Product Details