Gene Transcripts

Gene Symbol
LOC56764
Entrez Gene ID
56764
Gene Name
dnaj-like protein
Gene Aliases
Djl, rDJL
Chromosome Location
Chr. 2: 252500143 - 252505971 on Build Rnor_6.0
UniGene ID
Rn.92862
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020089.2 NP_064474.1 1 501
GenBank mRNA AF154849.1 - 1 475
BC083558.1 - 1 501 7134067

View Full Product Details