Gene Transcripts

Gene Symbol
Aadac
Entrez Gene ID
57300
Gene Name
arylacetamide deacetylase
Gene Aliases
Aada
Chromosome Location
Chr. 2: 150146234 - 150157480 on Build Rnor_6.0
UniGene ID
Rn.48732
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020538.1 NP_065413.1 1, 2 160
GenBank mRNA AF182426.1 - 1, 2 160
BC088143.1 - 1, 2 161 7323135

View Full Product Details