Gene Transcripts

Gene Symbol
Cep78
Entrez Gene ID
60347
Gene Name
centrosomal protein 78
Gene Aliases
Ip63
Chromosome Location
Chr. 1: 233185815 - 233214873 on Build Rnor_6.0
UniGene ID
Rn.42936
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021741.2 NP_068509.2 5 712
XM_006231136.2 XP_006231198.1 5 827
GenBank mRNA FQ222521.1 - 3 577
X99330.1 - 5 500

View Full Product Details