Gene Transcripts
- Gene Symbol
- Camk2b
- Entrez Gene ID
- 24245
- Gene Name
- calcium/calmodulin-dependent protein kinase II beta
- Gene Aliases
- Ck2b
- Chromosome Location
- Chr. 14: 86208876 - 86297751 on Build Rnor_6.0
- UniGene ID
- Rn.9743
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042354.1 |
NP_001035813.1
|
7, 8 |
597 |
|
NM_001042356.1 |
NP_001035815.1
|
7, 8 |
597 |
|
NM_021739.2 |
NP_068507.2
|
7, 8 |
597 |
|
XM_006251170.2 |
XP_006251232.1
|
7, 8 |
696 |
|
XM_006251171.2 |
XP_006251233.1
|
7, 8 |
696 |
|
XM_006251172.2 |
XP_006251234.1
|
7, 8 |
696 |
|
XM_006251174.2 |
XP_006251236.1
|
7, 8 |
696 |
|
XM_006251176.2 |
XP_006251238.1
|
7, 8 |
677 |
|
XM_006251177.2 |
XP_006251239.1
|
7, 8 |
676 |
|
GenBank mRNA |
M16112.1 |
- |
7, 8 |
577 |
|
X83375.1 |
- |
7, 8 |
559 |
|
View Full Product Details