Gene Transcripts

Gene Symbol
Unc5a
Entrez Gene ID
60629
Gene Name
unc-5 homolog A (C. elegans)
Gene Aliases
Unc5h1
Chromosome Location
Chr. 17: 10152949 - 10208599 on Build Rnor_6.0
UniGene ID
Rn.44433
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022206.1 NP_071542.1 2 260
XM_008771505.1 XP_008769727.1 2 499
GenBank mRNA U87305.1 - 2 260

View Full Product Details