Gene Transcripts

Gene Symbol
Slc26a1
Entrez Gene ID
64076
Gene Name
solute carrier family 26 (anion exchanger), member 1
Gene Aliases
sat-1
Chromosome Location
Chr. 14: 2050805 - 2056091 on Build Rnor_6.0
UniGene ID
Rn.10016
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022287.2 NP_071623.2 2 207
GenBank mRNA BC085735.1 - 2 207
L23413.1 - 2 529

View Full Product Details