Gene Transcripts

Gene Symbol
Slc5a2
Entrez Gene ID
64522
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 2
Gene Aliases
Sglt2
Chromosome Location
Chr. 1: 199682688 - 199688809 on Build Rnor_6.0
UniGene ID
Rn.5887
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022590.2 NP_072112.2 7 689
GenBank mRNA AB070849.1 - 6 824
AB070850.1 - 6 824
AB070976.1 - 7 684
U29881.1 - 7 689

View Full Product Details