Gene Transcripts

Gene Symbol
Pnpo
Entrez Gene ID
64533
Gene Name
pyridoxamine 5'-phosphate oxidase
Gene Aliases
Chromosome Location
Chr. 10: 84874926 - 84881190 on Build Rnor_6.0
UniGene ID
Rn.158883
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022601.2 NP_072123.1 2 344
GenBank mRNA BC087016.1 - 2 344
U91561.1 - 2 335

View Full Product Details