Gene Transcripts

Gene Symbol
Chrna9
Entrez Gene ID
65024
Gene Name
cholinergic receptor, nicotinic, alpha 9 (neuronal)
Gene Aliases
Chromosome Location
Chr. 14: 43883450 - 43890171 on Build Rnor_6.0
UniGene ID
Rn.54463
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022930.1 NP_075219.1 3 436
GenBank mRNA AY574257.1 - 3 349
U12336.1 - 3 436

View Full Product Details