Gene Transcripts

Gene Symbol
Kcns2
Entrez Gene ID
66022
Gene Name
potassium voltage-gated channel, modifier subfamily S, member 2
Gene Aliases
Chromosome Location
Chr. 7: 73559226 - 73590385 on Build Rnor_6.0
UniGene ID
Rn.64498
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023966.1 NP_076456.1 2 645
GenBank mRNA AJ296090.1 - 2 645

View Full Product Details