Gene Transcripts

Gene Symbol
OVOL1
Entrez Gene ID
5017
Gene Name
ovo like transcriptional repressor 1
Gene Aliases
HOVO1
Chromosome Location
Chr. 11: 65787034 - 65797219 on Build GRCh38
UniGene ID
Hs.134434
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004561.3 NP_004552.2 4 1816
XM_005274018.3 XP_005274075.1 4 1634
XM_011545067.2 XP_011543369.1 4 1632
XM_011545068.2 XP_011543370.1 4 2177
XM_017017837.1 XP_016873326.1 4 2470
XM_017017838.1 XP_016873327.1 4 1509
GenBank mRNA AF016045.1 - 3 1692
BC059408.1 - 4 1792 30343789
BX648725.1 - 4 1696

View Full Product Details