Gene Transcripts
- Gene Symbol
- Musk
- Entrez Gene ID
- 81725
- Gene Name
- muscle, skeletal, receptor tyrosine kinase
- Gene Aliases
- Nsk1
- Chromosome Location
- Chr. 5: 75392790 - 75498694 on Build Rnor_6.0
- UniGene ID
- Rn.10210
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031061.1 |
NP_112323.1
|
4 |
530 |
|
GenBank mRNA |
U34985.1 |
- |
4 |
530 |
|
View Full Product Details