Gene Transcripts

Gene Symbol
Amhr2
Entrez Gene ID
29530
Gene Name
anti-Mullerian hormone receptor, type II
Gene Aliases
Chromosome Location
Chr. 7: 144052202 - 144060678 on Build Rnor_6.0
UniGene ID
Rn.10165
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030998.1 NP_112260.1 5, 6 698
GenBank mRNA KJ923229.1 - 5, 6 698
U42427.1 - 5, 6 698
X71916.1 - 5, 6 680

View Full Product Details