Gene Transcripts

Gene Symbol
Rpl37
Entrez Gene ID
81770
Gene Name
ribosomal protein L37
Gene Aliases
Chromosome Location
Chr. 2: 54832634 - 54834587 on Build Rnor_6.0
UniGene ID
Rn.129486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031106.1 NP_112368.1 3 198
GenBank mRNA BC059132.1 - 3 195
BC069173.1 - 3 180 7103990
FQ210660.1 - 3 201
FQ210874.1 - 3 201
FQ217102.1 - 3 201
FQ217508.1 - 3 191
FQ220738.1 - 3 201
FQ221496.1 - 3 191
FQ221700.1 - 3 191
FQ222983.1 - 3 201
FQ223019.1 - 3 201
FQ223296.1 - 3 206
FQ223471.1 - 3 191
S79981.1 - 3 198
X66369.1 - 3 198

Gene Symbol
LOC100360841
Entrez Gene ID
100360841
Gene Name
ribosomal protein L37-like
Gene Aliases
Chromosome Location
Chr. 9: 69790835 - 69791192 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002730037.4 XP_002730083.1 1 194

Gene Symbol
RGD1561310
Entrez Gene ID
498744
Gene Name
similar to ribosomal protein L37
Gene Aliases
Chromosome Location
Chr. 17: 36438176 - 36438782 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771609.1 XP_008769831.1 1 194
XM_574022.5 XP_574022.3 1 194

View Full Product Details