Gene Transcripts
- Gene Symbol
- Rps27a
- Entrez Gene ID
- 81777
- Gene Name
- ribosomal protein S27a
- Gene Aliases
- Uba52, Ubb
- Chromosome Location
- Chr. 5: 138628482 - 138629026 on Build Rnor_6.0
- UniGene ID
- Rn.106034
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031113.1 |
NP_112375.1
|
1 |
239 |
|
- Gene Symbol
- LOC100912032
- Entrez Gene ID
- 100912032
- Gene Name
- ubiquitin-40S ribosomal protein S27a-like
- Gene Aliases
- Chromosome Location
- Chr. 14: 113966573 - 113968469 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC681260
- Entrez Gene ID
- 681260
- Gene Name
- similar to ribosomal protein S27a
- Gene Aliases
- Chromosome Location
- Chr. 1: 255572806 - 255573402 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- Rps27l3
- Entrez Gene ID
- 498837
- Gene Name
- ribosomal protein S27-like 3
- Gene Aliases
- Gm8430, RGD1564290
- Chromosome Location
- Chr. 18: 25394170 - 25394839 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details