Gene Transcripts

Gene Symbol
Cd8b
Entrez Gene ID
24931
Gene Name
CD8b molecule
Gene Aliases
Cd8b1
Chromosome Location
Chr. 4: 99185960 - 99201887 on Build Rnor_6.0
UniGene ID
Rn.10330
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031539.1 NP_113727.1 2 224
GenBank mRNA BC059127.1 - 2 246
X04310.1 - 2 224

View Full Product Details