Gene Transcripts
- Gene Symbol
- Dio2
- Entrez Gene ID
- 65162
- Gene Name
- deiodinase, iodothyronine, type II
- Gene Aliases
- 5DII, DIOII
- Chromosome Location
- Chr. 6: 114479638 - 114489443 on Build Rnor_6.0
- UniGene ID
- Rn.88380
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031720.3 |
NP_113908.3
|
1 |
746 |
|
GenBank mRNA |
U53505.1 |
- |
1 |
746 |
|
View Full Product Details