Gene Transcripts
- Gene Symbol
- Clic4
- Entrez Gene ID
- 83718
- Gene Name
- chloride intracellular channel 4
- Gene Aliases
- Chromosome Location
- Chr. 5: 153568937 - 153625669 on Build Rnor_6.0
- UniGene ID
- Rn.104756
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031818.1 |
NP_114006.1
|
2, 3 |
190 |
|
GenBank mRNA |
AF104119.1 |
- |
2, 3 |
190 |
|
View Full Product Details