Gene Transcripts

Gene Symbol
Chrnb4
Entrez Gene ID
25103
Gene Name
cholinergic receptor, nicotinic, beta 4 (neuronal)
Gene Aliases
Chromosome Location
Chr. 8: 59610489 - 59629073 on Build Rnor_6.0
UniGene ID
Rn.9695
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052806.2 NP_434693.1 2 86
GenBank mRNA AY574260.1 - 2 86
U42976.1 - 146
X15834.1 - 198

View Full Product Details