Gene Transcripts

Gene Symbol
Nr1i2
Entrez Gene ID
84385
Gene Name
nuclear receptor subfamily 1, group I, member 2
Gene Aliases
PXR
Chromosome Location
Chr. 11: 65022100 - 65058546 on Build Rnor_6.0
UniGene ID
Rn.207164
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052980.2 NP_443212.1 2 352
GenBank mRNA AF151377.1 - 2 325
BC091138.1 - 2 353
BC107449.1 - 1 114
CO575646.1 - 2 352

View Full Product Details