Gene Transcripts

Gene Symbol
Pgam1
Entrez Gene ID
24642
Gene Name
phosphoglycerate mutase 1 (brain)
Gene Aliases
Pgmut
Chromosome Location
Chr. 1: 261158261 - 261165808 on Build Rnor_6.0
UniGene ID
Rn.1383
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053290.1 NP_445742.1 2 430
GenBank mRNA BC065582.1 - 2 485
M76591.1 - 2 430

View Full Product Details