Gene Transcripts

Gene Symbol
Hs3st1
Entrez Gene ID
84406
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 1
Gene Aliases
Chromosome Location
Chr. 14: 75880410 - 75881346 on Build Rnor_6.0
UniGene ID
Rn.17364
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053391.1 NP_445843.1 1 150
GenBank mRNA AF177430.1 - 1 150

View Full Product Details