Gene Transcripts

Gene Symbol
Snx1
Entrez Gene ID
84471
Gene Name
sorting nexin 1
Gene Aliases
Chromosome Location
Chr. 8: 71745687 - 71786182 on Build Rnor_6.0
UniGene ID
Rn.144614
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053411.1 NP_445863.1 3 413
XM_006243317.2 XP_006243379.1 3 296
XM_008766349.1 XP_008764571.1 3 468
GenBank mRNA AF218916.1 - 3 413
BC092201.1 - 3 416 7309531

View Full Product Details