Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053437.1 | NP_445889.1 | 4 | 427 | |
GenBank mRNA | AB062759.1 | - | 4 | 405 | |
AB062760.1 | - | 4 | 405 | ||
AB062761.1 | - | 4 | 405 | ||
AB062762.1 | - | 4 | 405 | ||
AB062763.1 | - | 4 | 403 | ||
AF296131.1 | - | 4 | 427 | ||
BC081742.1 | - | 4 | 399 |