Gene Transcripts

Gene Symbol
Tekt1
Entrez Gene ID
85270
Gene Name
tektin 1
Gene Aliases
Chromosome Location
Chr. 10: 58895689 - 58924098 on Build Rnor_6.0
UniGene ID
Rn.46442
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053508.2 NP_445960.1 3 297
GenBank mRNA AJ306427.1 - 2 226
BC078773.1 - 3 297 7133616

View Full Product Details