Gene Transcripts

Gene Symbol
Prdx6
Entrez Gene ID
94167
Gene Name
peroxiredoxin 6
Gene Aliases
Chromosome Location
Chr. 13: 79077567 - 79088113 on Build Rnor_6.0
UniGene ID
Rn.42
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053576.2 NP_446028.1 2 219
GenBank mRNA AF014009.1 - 2 219
FQ218168.1 - 2 248
Y17295.1 - 233

View Full Product Details