Gene Transcripts
- Gene Symbol
- Ankh
- Entrez Gene ID
- 114506
- Gene Name
- ANKH inorganic pyrophosphate transport regulator
- Gene Aliases
- Ank
- Chromosome Location
- Chr. 2: 80131563 - 80256948 on Build Rnor_6.0
- UniGene ID
- Rn.211841
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053714.1 |
NP_446166.1
|
4, 5 |
827 |
|
GenBank mRNA |
AF393241.1 |
- |
4, 5 |
827 |
|
View Full Product Details