Gene Transcripts

Gene Symbol
Spata2
Entrez Gene ID
114210
Gene Name
spermatogenesis associated 2
Gene Aliases
Chromosome Location
Chr. 3: 164229211 - 164239494 on Build Rnor_6.0
UniGene ID
Rn.201291
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053675.2 NP_446127.3 3 853
XM_006235636.2 XP_006235698.1 3 764
GenBank mRNA AF123651.1 - 2 654
BC081752.1 - 3 853 7134513

View Full Product Details