Gene Transcripts

Gene Symbol
Rraga
Entrez Gene ID
117044
Gene Name
Ras-related GTP binding A
Gene Aliases
Raga
Chromosome Location
Chr. 5: 104941067 - 104942662 on Build Rnor_6.0
UniGene ID
Rn.2816
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053973.2 NP_446425.1 1 811
GenBank mRNA BC061850.1 - 1 812
X85183.1 - 1 796

View Full Product Details