Gene Transcripts
- Gene Symbol
- Slc8a2
- Entrez Gene ID
- 140447
- Gene Name
- solute carrier family 8 (sodium/calcium exchanger), member 2
- Gene Aliases
- Ncx2
- Chromosome Location
- Chr. 1: 78029555 - 78054042 on Build Rnor_6.0
- UniGene ID
- Rn.10305
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_078619.1 |
NP_511174.1
|
6 |
2000 |
|
GenBank mRNA |
U08141.1 |
- |
6 |
2000 |
|
View Full Product Details