Gene Transcripts

Gene Symbol
ZKSCAN5
Entrez Gene ID
23660
Gene Name
zinc finger with KRAB and SCAN domains 5
Gene Aliases
ZFP-95, ZFP95, ZNF914, ZSCAN37
Chromosome Location
Chr. 7: 99501296 - 99534106 on Build GRCh38
UniGene ID
Hs.110839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318082.1 NP_001305011.1 2 287
NM_001318083.1 NP_001305012.1 2 225
NM_001318084.1 NP_001305013.1 1 135
NM_014569.3 NP_055384.1 2 346
NM_145102.3 NP_659570.1 2 225
XM_017011918.1 XP_016867407.1 4 684
XM_017011919.1 XP_016867408.1 3 727
XM_017011920.1 XP_016867409.1 1 892
XM_017011921.1 XP_016867410.1 2 658
XM_017011922.1 XP_016867411.1 2 317
XM_017011923.1 XP_016867412.1 4 684
XM_017011924.1 XP_016867413.1 2 346
XM_017011926.1 XP_016867415.1 2 385
XM_017011927.1 XP_016867416.1 2 317
GenBank mRNA AB023232.1 - 2 287
AF170025.1 - 2 199
AK027714.1 - 2 315
AK096552.1 - 2 884
BC030790.2 - 2 256 4827870
BX648490.1 - 1 888
CX163381.1 - 2 192
DA044307.1 - 2 334
DA572286.1 - 2 288
DA580238.1 - 2 219
DA773579.1 - 2 346

View Full Product Details