Gene Transcripts

Gene Symbol
PPM1E
Entrez Gene ID
22843
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1E
Gene Aliases
CaMKP-N, POPX1, PP2CH, caMKN
Chromosome Location
Chr. 17: 58755869 - 58985617 on Build GRCh38
UniGene ID
Hs.245044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014906.4 NP_055721.3 2, 3 711
XM_017024359.1 XP_016879848.1 2, 3 1228
GenBank mRNA AB028995.1 - 2, 3 715
AF260269.1 - 2, 3 742
AF520614.1 - 2, 3 588
AK289966.1 - 2, 3 717
BC136290.1 - 2, 3 657 9020277
BC151228.1 - 2, 3 715 9007247

View Full Product Details