Gene Transcripts
- Gene Symbol
- Gzmc
- Entrez Gene ID
- 171290
- Gene Name
- granzyme C
- Gene Aliases
- Rnpk-4, Rnpk4
- Chromosome Location
- Chr. 15: 35392158 - 35394787 on Build Rnor_6.0
- UniGene ID
- Rn.88335
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134332.2 |
NP_599159.1
|
2, 3 |
218 |
|
GenBank mRNA |
FQ219852.1 |
- |
2, 3 |
281 |
|
FQ219889.1 |
- |
2, 3 |
281 |
|
FQ219966.1 |
- |
2, 3 |
279 |
|
FQ219998.1 |
- |
2, 3 |
283 |
|
FQ220005.1 |
- |
2, 3 |
281 |
|
FQ220014.1 |
- |
2, 3 |
281 |
|
FQ220035.1 |
- |
2, 3 |
281 |
|
FQ220062.1 |
- |
2, 3 |
283 |
|
FQ220064.1 |
- |
2, 3 |
281 |
|
FQ220079.1 |
- |
2, 3 |
279 |
|
FQ220128.1 |
- |
2, 3 |
279 |
|
FQ220142.1 |
- |
2, 3 |
281 |
|
FQ220223.1 |
- |
2, 3 |
281 |
|
FQ220228.1 |
- |
2, 3 |
271 |
|
FQ220350.1 |
- |
2, 3 |
281 |
|
FQ220364.1 |
- |
2, 3 |
281 |
|
FQ220426.1 |
- |
2, 3 |
281 |
|
FQ220453.1 |
- |
2, 3 |
281 |
|
FQ220464.1 |
- |
2, 3 |
281 |
|
FQ220466.1 |
- |
2, 3 |
281 |
|
FQ220480.1 |
- |
2, 3 |
283 |
|
FQ220512.1 |
- |
2, 3 |
281 |
|
FQ220556.1 |
- |
2, 3 |
281 |
|
FQ220562.1 |
- |
2, 3 |
281 |
|
FQ220600.1 |
- |
2, 3 |
281 |
|
FQ220618.1 |
- |
2, 3 |
281 |
|
FQ220735.1 |
- |
2, 3 |
281 |
|
FQ220749.1 |
- |
2, 3 |
280 |
|
FQ220751.1 |
- |
2, 3 |
279 |
|
FQ220856.1 |
- |
2, 3 |
280 |
|
FQ220878.1 |
- |
2, 3 |
280 |
|
FQ220928.1 |
- |
2, 3 |
283 |
|
FQ221390.1 |
- |
2, 3 |
281 |
|
FQ221473.1 |
- |
2, 3 |
281 |
|
FQ221940.1 |
- |
2, 3 |
281 |
|
FQ223110.1 |
- |
2, 3 |
281 |
|
FQ223477.1 |
- |
2, 3 |
280 |
|
FQ228922.1 |
- |
2, 3 |
283 |
|
FQ229160.1 |
- |
2, 3 |
281 |
|
FQ229161.1 |
- |
2, 3 |
281 |
|
FQ229162.1 |
- |
2, 3 |
281 |
|
FQ229181.1 |
- |
2, 3 |
283 |
|
FQ229215.1 |
- |
2, 3 |
281 |
|
FQ229235.1 |
- |
2, 3 |
285 |
|
FQ229289.1 |
- |
2, 3 |
281 |
|
FQ229298.1 |
- |
2, 3 |
281 |
|
FQ229311.1 |
- |
2, 3 |
281 |
|
FQ229314.1 |
- |
2, 3 |
281 |
|
FQ229333.1 |
- |
2, 3 |
281 |
|
FQ229395.1 |
- |
2, 3 |
281 |
|
FQ229439.1 |
- |
2, 3 |
279 |
|
FQ229452.1 |
- |
2, 3 |
281 |
|
FQ229454.1 |
- |
2, 3 |
281 |
|
FQ229468.1 |
- |
2, 3 |
281 |
|
FQ229486.1 |
- |
2, 3 |
281 |
|
FQ229502.1 |
- |
2, 3 |
281 |
|
FQ229509.1 |
- |
2, 3 |
281 |
|
FQ229543.1 |
- |
2, 3 |
281 |
|
FQ229636.1 |
- |
2, 3 |
281 |
|
FQ229685.1 |
- |
2, 3 |
281 |
|
FQ229690.1 |
- |
2, 3 |
325 |
|
FQ229764.1 |
- |
2, 3 |
282 |
|
FQ229772.1 |
- |
2, 3 |
279 |
|
FQ229825.1 |
- |
2, 3 |
281 |
|
FQ229868.1 |
- |
2, 3 |
312 |
|
FQ229912.1 |
- |
2, 3 |
281 |
|
FQ229913.1 |
- |
2, 3 |
279 |
|
FQ229994.1 |
- |
2, 3 |
283 |
|
FQ230028.1 |
- |
2, 3 |
283 |
|
FQ230087.1 |
- |
2, 3 |
281 |
|
FQ230100.1 |
- |
2, 3 |
281 |
|
FQ230157.1 |
- |
2, 3 |
283 |
|
FQ230165.1 |
- |
2, 3 |
281 |
|
U57062.1 |
- |
2, 3 |
219 |
|
- Gene Symbol
- LOC100911163
- Entrez Gene ID
- 100911163
- Gene Name
- granzyme C-like
- Gene Aliases
- Chromosome Location
- Chr. 15: 36247356 - 36252913 on Build Rnor_6.0
- Species
- Rattus norvegicus
- Gene Symbol
- LOC691695
- Entrez Gene ID
- 691695
- Gene Name
- similar to granzyme C
- Gene Aliases
- Chromosome Location
- Chr. 15: 36448970 - 36451720 on Build Rnor_6.0
- UniGene ID
- Rn.88335
- Species
- Rattus norvegicus
View Full Product Details