Gene Transcripts

Gene Symbol
Giot1
Entrez Gene ID
171090
Gene Name
gonadotropin inducible ovarian transcription factor 1
Gene Aliases
Chromosome Location
Chr. 7: 13104179 - 13122614 on Build Rnor_6.0
UniGene ID
Rn.107259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133563.1 NP_598247.1 4 835
GenBank mRNA AB047636.1 - 4 835

View Full Product Details