Gene Transcripts

Gene Symbol
Taar1
Entrez Gene ID
113914
Gene Name
trace-amine-associated receptor 1
Gene Aliases
Tar1, Trar1, taR-1
Chromosome Location
Chr. 1: 22595477 - 22596475 on Build Rnor_6.0
UniGene ID
Rn.81209
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134328.1 NP_599155.1 1 512
GenBank mRNA AF421352.1 - 1 512

View Full Product Details